Få mere at vide
Brady™ S3100 Sign and Label Printer
Artikelnummer. 31842975
Skift visning
Tastaturtype:
AZERTY
QWERTY
QWERTZ
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Tastaturtype
31842975 AZERTY
31842972 QWERTY
31842971 QWERTZ
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 31842975 Leverandør Brady™ Leverandørnr. 149517
The easy-to-use Brady Printer S3100 sign and label printer is designed to meet a variety of your facility identification needs, helping you save time so you can get back to work.
  • Create the labels you need, when you need them without the wait with do-it-yourself label creation
  • Ideal for multi-user areas in your facility, anyone can simply walk up, create labels and print right on the spot
  • The built-in keyboard and touchscreen interface make label creation a breeze
  • Label creation is even faster with the quick setup and print speeds that come with print smart technology
  • Prints on a variety of material options so you can create signs and labels for a variety of applications
  • Ships with free Brady Workstation Basic Label Design software suite, which will enable you to immediately design professional labels.
Tekniske data
Produkttype Printer
Farve Blue, Gray
Display Type Color LCD Touchscreen
Til brug med (udstyr) B30 Series Thermal Transfer Tapes and Ribbons, B30 Series Thermal Transfer Tapes
Materiale Plastic
Print teknologi Thermal Transfer
Længde (engelsk) 10.25 in.
Bredde (engelsk) 12 in.
Længde (metrisk) 260.35 mm
Bredde (metrisk) 304.80 mm
Højde (engelsk) 9.5 in.
Højde (metrisk) 241.30 mm
Dimensioner (L x B x H) 260.35 x 304.80 x 241.30 mm
Autoklaverbar Not Autoclavable
Sterilitet Non-sterile
Farveevne Monocolour
Skærmstørrelse 155.44 x 91.95 mm
Inkluderer S3100 Printer, Power Cord, USB Cable, Cleaning Kit, Cutter Cleaning Tool, Quick Start Guide, Brady Workstation Basic Suite, Stylus and Mouse
Sprog Bulgarian, Chinese (Simplified), Chinese (Traditional), Croatian, Czech, Danish, Dutch, English(US), Estonian, Finnish, French, German, Hungarian, Italian, Korean, Norwegian, Polish, Portuguese, Romanian, Russian, Slovak, Slovenian, Spanish
Hukommelse 256 MB
Model- S3100-AZ
Udskrivningshastighed 76.00 mm/sec.
Opløsning 300 dpi
Konnektortyper Ethernet, USB
Vægt (engelsk) 20 lb.
Vægt (metrisk) 9.07 kg
Certificeringer/compliance UL, cUL, CE, WEEE, RoHS, RCM (Australia/New Zealand), EAC (Russia, Belarus, and Kazakhstan), China: CCC-RoHS
Til brug med (applikation) Asset tracking, Cold temperature labelling, Component and equipment labelling, Electrical labelling, Exit and egress marking, Facility identification, Floor marking, General identification, Hazard communication identification, Lean and 5S labelling, Lockout tagout, Pipe and valve marking, Safety identification, Temperature indicating labelling
Mængde 1 Ea.
garanti 1 Year
Spænding 110/240 V
Tastaturtype AZERTY
Alternativt varenummer S3100-AZ
Stiktype AC Adaptor