Få mere at vide
Tygon S3™ E-3603 Vakuumslange
Shop alle Cole-Parmer produkter
Diameter (engelsk) Indre:
1 tomme
1/2 tomme
1/4 tomme
3/16 tommer
3/4 tommer
3/8 tommer
5/8 tommer
Diameter (engelsk) Ydre:
1-1/2 in.
1-1/8 tommer
1-3/8 tommer
2''
5/8 tommer
7/8 tommer
9/16 tommer
Beskrivelse
Bio-based formulation reduces environmental impact. Lot-to-lot consistency for reproducible results. Easily slips over fittings and grips securely. Contains no BPA or phthalates. Meets all USP Class VI and FDA food contact criteria. . Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Tekniske data
Tekniske data
| Certificeringer/compliance | USP;FDA |
| Diameter (engelsk) Indre | 1/2 tomme |
| Materiale | Tygon S3 E-3603 |
| Diameter (engelsk) Ydre | 1-1/8 tommer |
| Steriliseringsmetode | EtO;Autoclave |