missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
S1P3/EDG-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-14467
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
S1P3/EDG-3 Polyclonal antibody specifically detects S1P3/EDG-3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Tekniske data
| S1P3/EDG-3 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EDG-3, EDG3FLJ37523, Endothelial differentiation G-protein coupled receptor 3, endothelial differentiation, sphingolipid G-protein-coupled receptor, 3, FLJ93220, G protein-coupled receptor, endothelial differentiation gene-3, LPB3, MGC71696, S1P receptor 3, S1P receptor Edg-3, S1P receptor EDG3, S1P3, sphingosine 1-phosphate receptor 3, Sphingosine 1-phosphate receptor Edg-3, sphingosine-1-phosphate receptor 3 | |
| Synthetic 16 amino acid peptide from N-Terminus extracellular domain of human S1P3/EDG-3. | |
| 50 μg | |
| Cell Biology, Cell Cycle and Replication, Cellular Markers, GPCR, Lipid and Metabolism, Signal Transduction | |
| 1903 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion