missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
S100A8 Antibody (CL11171) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07985AF700
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
S100A8 Monoclonal antibody specifically detects S100A8 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Tekniske data
| S100A8 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 60B8AG, CAGACP-10, calgranulin A, calgranulin-A, Calprotectin L1L subunit, CFAGL1Ag, CGLA, Cystic fibrosis antigen, Leukocyte L1 complex light chain, MA387, MIF, Migration inhibitory factor-related protein 8, MRP-8, MRP8S100 calcium binding protein A8 (calgranulin A), NIF, P8, protein S100-A8, S100 calcium binding protein A8, S100 calcium-binding protein A8, S100 calcium-binding protein A8 (calgranulin A), Urinary stone protein band A | |
| This antibody was developed using a recombinant protein derived from P05109, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer | |
| 6279 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL11171 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion