Få mere at vide
(S)-Tetrahydro-Alpha-(1-methylethyl)-2-oxo-1(2H)-pyrimidineacetic Acid-d8, TRC
Artikelnummer. 30423191
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30423191 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30423191 Leverandør Toronto Research Chemicals Leverandørnr. TRCT293902100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-Tetrahydro-alpha-(1-methylethyl)-2-oxo-1(2H)-pyrimidineacetic Acid-d8
CAS 1323257-58-4
Molekylær formel C9H8D8N2O3
Mængde 100 mg
Synonym (alphaS)-Tetrahydro-alpha-(1-methylethyl)-2-oxo-1(2H)-pyrimidineacetic Acid-d8
InChI formel InChI=1S/C9H16N2O3/c1-6(2)7(8(12)13)11-5-3-4-10-9(11)14/h6-7H,3-5H2,1-2H3,(H,10,14)(H,12,13)/i1D3,2D3,6D,7D
SMIL [2H]C([2H])([2H])C([2H])(C([2H])([2H])[2H])C([2H])(N1CCCNC1=O)C(=O)O
IUPAC navn 2,3,4,4,4-pentadeuterio-2-(2-oxo-1,3-diazinan-1-yl)-3-(trideuteriomethyl)butanoic acid
Molekylvægt (g/mol) 208.28
Formel vægt 208.1663