Få mere at vide
(S)-Silodosin, TRC
Artikelnummer. 30010166
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30010166 2.5 mg 2.5mg
30007396 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30010166 Leverandør Toronto Research Chemicals Leverandørnr. TRCS4650052.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2182279-45-2
Molekylær formel C25H32F3N3O4
Molekylvægt (g/mol) 495.53
Synonym Silodosin Impurity 4
IUPAC navn 1-(3-hydroxypropyl)-5-[(2S)-2-[2-[2-(2,2,2-trifluoroethoxy)phenoxy]ethylamino]propyl]indoline-7-carboxamide
SMIL C[C@@H](Cc1cc2CCN(CCCO)c2c(c1)C(=O)N)NCCOc3ccccc3OCC(F)(F)F
Tekniske data
Kemisk navn eller materiale (S)-Silodosin
Mængde 2.5 mg
InChI formel InChI=1S/C25H32F3N3O4/c1-17(30-8-12-34-21-5-2-3-6-22(21)35-16-25(26,27)28)13-18-14-19-7-10-31(9-4-11-32)23(19)20(15-18)24(29)33/h2-3,5-6,14-15,17,30,32H,4,7-13,16H2,1H3,(H2,29,33)/t17-/m0/s1
Formel vægt 495.2345