Learn More
Fisherbrand™ S-Series Heated Ultrasonic Cleaning Bath
With state of the art microprocessor controlled ultrasonic cleaning and sweep technology. Fisherbrand™ S-Series Ultrasonic Cleaners offer a comprehensive range of analogue ultrasonic water baths covering the requirements for laboratory research, sonochemistry and degassing applications.
Brand:  Fisherbrand™ 1003946
 View more versions of this product
Product Code. 10427912
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Long lifespan from cavitation resistant stainless steel tanks
  • LED display showing set and remaining time and pre-set and actual temperature
  • Ergonomically shaped plastic carrying handles
  • Newly developed cover reduces noise and accelerates the heating up process
  • Easy draining of cleaning liquid though the drain duct via a dial on the side
  • Operating panel arranged so that liquid cannot enter the electronics
  • Degas function key
Specifications
Specifications
V2A Stainless Steel Casing, V4A Stainless Steel Tank
90 L
230VAC, 2800w
Elmasonic S 900 (H)
42 kg
230 VAC
570 x 715 x 467 mm
Laboratory, medical sector, dental laboratory, jewellery, watch sector, optician, workshop and service, industry
30°C to 80°C
Documents
One moment while we fetch your results.
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ S-Series Heated Ultrasonic Cleaning Bath > Capacity: 90L; With Heating