Få mere at vide
(S)-Scoulerine, TRC
Artikelnummer. 30011428
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011428 5 mg
30004692 10 mg
30005847 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011428 Leverandør Toronto Research Chemicals Leverandørnr. TRCS2008005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6451-73-6
Molekylær formel C19 H21 N O4
Molekylvægt (g/mol) 327.37
Synonym 6H-Dibenzo[a,g]quinolizine-2,9-diol, 5,8,13,13a-tetrahydro-3,10-dimethoxy-, (13aS)-, 13aα-Berbine-2,9-diol, 3,10-dimethoxy- (8CI), 6H-Dibenzo[a,g]quinolizine-2,9-diol, 5,8,13,13a-tetrahydro-3,10-dimethoxy-, (S)-, Scoulerine (7CI), (13aS)-5,8,13,13a-Tetrahydro-3,10-dimethoxy-6H-dibenzo[a,g]quinolizine-2,9-diol, (-)-Scoulerine, (13aS)-3,10-Dimethoxy-6,8,13,13a-tetrahydro-5H-isoquinolino[2,1-b]isoquinoline-2,9-diol, (S)-(-)-Scoulerine, (S)-Scoulerine, Alkaloid HF 1, from Hunnemannia fumariaefolia, Scoulerin, Scoulerine, (-)-, l-Scoulerine
IUPAC navn (13aS)-3,10-dimethoxy-6,8,13,13a-tetrahydro-5H-isoquinolino[3,2-a]isoquinoline-2,9-diol
SMIL COc1cc2CCN3Cc4c(C[C@H]3c2cc1O)ccc(OC)c4O
Tekniske data
Kemisk navn eller materiale Scoulerin
Renhedsgrad noter HPLC
Mængde 5 mg
InChI formel InChI=1S/C19H21NO4/c1-23-17-4-3-11-7-15-13-9-16(21)18(24-2)8-12(13)5-6-20(15)10-14(11)19(17)22/h3-4,8-9,15,21-22H,5-7,10H2,1-2H3/t15-/m0/s1
Formel vægt 327.1471
Procent renhed >95%