Få mere at vide
(S,S)-Palonosetron-d3 Hydrochloride, TRC
Artikelnummer. 30416784
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30416784 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30416784 Leverandør Toronto Research Chemicals Leverandørnr. TRCP16580210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S,S)-Palonosetron-d3 Hydrochloride
CAS 1246816-81-8
Renhedsgrad noter HPLC
Molekylær formel C19H22D3ClN2O
Mængde 10 mg
Synonym (3aS)-2-(3S)-1-Azabicyclo[2.2.2]oct-3-yl-2,3,3a,4,5,6-hexahydro-1H-benz[de]isoquinolin-1-one-d3 Hydrochloride, Aloxi-d3, Onicit-d3, RS-25259-197-d3
InChI formel InChI=1S/C19H24N2O.ClH/c22-19-16-6-2-4-14-3-1-5-15(18(14)16)11-21(19)17-12-20-9-7-13(17)8-10-20;/h2,4,6,13,15,17H,1,3,5,7-12H2;1H/t15-,17-;/m1./s1/i11D2,17D;
SMIL O=C(N([C@@]1([2H])[C@@H](CC2)CC[N@@]2C1)C3([2H])[2H])C4=CC=CC5=C4[C@]3([H])CCC5.Cl
IUPAC navn (3aS)-3,3-dideuterio-2-[(3S)-3-deuterio-1-azabicyclo[2.2.2]octan-3-yl]-3a,4,5,6-tetrahydrobenzo[de]isoquinolin-1-one;hydrochloride
Molekylvægt (g/mol) 335.89
Formel vægt 335.1844
Procent renhed >95