Få mere at vide
(S)-Rivastigmine, TRC
Artikelnummer. 30135474
Skift visning
Mængde:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135474 1 g
30012237 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135474 Leverandør Toronto Research Chemicals Leverandørnr. TRCR5409951G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 123441-03-2
Molekylær formel C14 H22 N2 O2
Molekylvægt (g/mol) 250.34
Synonym Rivastigmine, 3-[(1S)-1-(Dimethylamino)ethyl]phenyl ethyl(methyl)carbamate, Carbamic acid, N-ethyl-N-methyl-, 3-[(1S)-1-(dimethylamino)ethyl]phenyl ester, Carbamic acid, ethylmethyl-, 3-[(1S)-1-(dimethylamino)ethyl]phenyl ester (9CI), Carbamic acid, ethylmethyl-, 3-[1-(dimethylamino)ethyl]phenyl ester, (S)-, ENA 713 free base, Exelon, Prometax, Rivastach, S-Rivastigmine, SDZ 212-713, [3-[(1S)-1-(Dimethylamino)ethyl]phenyl] N-ethyl-N-methylcarbamate
IUPAC navn [3-[(1S)-1-(dimethylamino)ethyl]phenyl] N-ethyl-N-methylcarbamate
SMIL CCN(C)C(=O)Oc1cccc(c1)[C@H](C)N(C)C
Tekniske data
Kemisk navn eller materiale Rivastigmine
Mængde 1 g
InChI formel InChI=1S/C14H22N2O2/c1-6-16(5)14(17)18-13-9-7-8-12(10-13)11(2)15(3)4/h7-11H,6H2,1-5H3/t11-/m0/s1
Formel vægt 250.1681