Få mere at vide
(S)-(+)-Repaglinide-d5 (ethoxy-d5), CDN
Artikelnummer. 30324222
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30324222 0.005 g
30321199 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324222 Leverandør CDN Leverandørnr. CDND71240.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Repaglinide-ethyl-d5
CAS 1217709-85-7
Molekylær formel C27 2H5 H31 N2 O4
Mængde 0.005 g
InChI formel InChI=1 S/C27H36N2O4/c1-4-33-25-17-20(12-13-22(25)27(31)32)18-26(30)28-23(16-19(2)3)21-10-6-7-11-24(21)29-14-8-5-9-15-29/h6-7,10-13,17,19,23 H,4-5,8-9,14-16,18H2,1-3H3,(H,28,30)(H,31,32)/t23-/m0/s1/i1D3,4D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])Oc1cc(CC(=O)N[C@@H](CC(C)C)c2ccccc2N3CCCCC3)ccc1C(=O)O
IUPAC navn 4-[2-[[(1 S)-3-methyl-1-(2-piperidin-1-ylphenyl)butyl]amino]-2-oxoethyl]-2-(1,1,2,2,2-pentadeuterioethoxy)benzoic acid
Molekylvægt (g/mol) 457.62
Formel vægt 457.2989 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity