Få mere at vide
(S)-Rasagiline Mesylate, TRC
Artikelnummer. 30133696
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133696 5 mg
30011276 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133696 Leverandør Toronto Research Chemicals Leverandørnr. TRCR1260055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 202464-88-8
Molekylær formel C13H17NO3S
Molekylvægt (g/mol) 267.34
Synonym (1S)-2,3-Dihydro-N-2-propyn-1-yl-1H-inden-1-amine Mesylate, (S)-2,3-Dihydro-N-2-propynyl-1H-inden-1-amine Mesylate, TVP 1022 Mesylate;
IUPAC navn (S)-N-(Prop-2-yn-1-yl)-2,3-dihydro-1H-inden-1-amine Methanesulfonate
SMIL CS(=O)(=O)O.C#CCN[C@H]1CCC2=CC=CC=C12
Tekniske data
Kemisk navn eller materiale (S)-Rasagiline Mesylate
Renhedsgrad noter HPLC
Mængde 5 mg
InChI formel InChI=1S/C12H13N.CH4O3S/c1-2-9-13-12-8-7-10-5-3-4-6-11(10)12;1-5(2,3)4/h1,3-6,12-13H,7-9H2;1H3,(H,2,3,4)/t12-;/m0./s1
Formel vægt 267.0929
Procent renhed >95%