Få mere at vide
MedChemExpress (S,R,S)-AHPC-PEG2-NH2 hydrochloride
Tekniske data
Tekniske data
| Molekylvægt (g/mol) | 612.18 g/mol |
| CAS | 2097973-72-1 |
| Indhold og opbevaring | 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture) |
| Forsendelsestilstand | Room temperature |
| Mængde | 100 mg |
| Synonym | VH032-PEG2-NH2 hydrochloride ∣ VHL Ligand-Linker Conjugates 3 hydrochloride ∣ E3 ligase Ligand-Linker Conjugates 6 |
| Opløselighed | H2O : 100 mg/mL (163.35 mM; Need ultrasonic) ∣ DMSO : 100 mg/mL (163.35 mM; Need ultrasonic) |