Få mere at vide
(S,R)-N-(1-Phenylethyl) Ibuprofen Amide, TRC
Artikelnummer. 30427072
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30427072 5 mg
30424411 25 mg
30421795 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30427072 Leverandør Toronto Research Chemicals Leverandørnr. TRCP3208955MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 81576-47-8
Molekylær formel C21H27NO
Molekylvægt (g/mol) 309.45
Synonym (αR)-α-Methyl-4-(2-methylpropyl)-N-[(1S)-1-phenylethyl]benzeneacetamide, [S-(R*,S*)]-α-Methyl-4-(2-methylpropyl)-N-(1-phenylethyl)benzeneacetamide
IUPAC navn (2R)-2-[4-(2-methylpropyl)phenyl]-N-[(1S)-1-phenylethyl]propanamide
SMIL C[C@H](C1=CC=C(C=C1)CC(C)C)C(=O)N[C@@H](C)C2=CC=CC=C2
Tekniske data
Kemisk navn eller materiale (S,R)-N-(1-Phenylethyl) Ibuprofen Amide
Mængde 5 mg
InChI formel InChI=1S/C21H27NO/c1-15(2)14-18-10-12-19(13-11-18)16(3)21(23)22-17(4)20-8-6-5-7-9-20/h5-13,15-17H,14H2,1-4H3,(H,22,23)/t16-,17+/m1/s1
Formel vægt 309.2093