Få mere at vide
(S)-Pulegone, TRC
Artikelnummer. 30426389
Skift visning
Mængde:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30426389 5 mL
30409753 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30426389 Leverandør Toronto Research Chemicals Leverandørnr. TRCP8401705ML
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale S-(-)-Pulegone
CAS 3391-90-0
Molekylær formel C10 H16 O
Mængde 5 mL
Synonym (S)-5-methyl-2-(1-methylethylidene)-cyclohexane, (S)-(-)-p-Menth-4(8)-en-3-one
InChI formel InChI=1S/C10H16O/c1-7(2)9-5-4-8(3)6-10(9)11/h8H,4-6H2,1-3H3/t8-/m0/s1
SMIL C[C@H]1CCC(=C(C)C)C(=O)C1
IUPAC navn (5S)-5-methyl-2-propan-2-ylidenecyclohexan-1-one
Molekylvægt (g/mol) 152.233
Formel vægt 152.12