Få mere at vide
(S)-Propyl Pramipexole, TRC
Artikelnummer. 30138323
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138323 100 mg
30137466 50 mg
30004480 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138323 Leverandør Toronto Research Chemicals Leverandørnr. TRCT293690100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1147937-31-2
Molekylær formel C13 H23 N3 S
Molekylvægt (g/mol) 253.41
Synonym (S)-N6,N6-Dipropyl-4,5,6,7-tetrahydrobenzo[d]thiazole-2,6-diamine, (6S)-4,5,6,7-Tetrahydro-N6,N6-dipropyl-2,6-benzothiazolediamine
IUPAC navn (6S)-N6,N6-dipropyl-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine
SMIL CCCN(CCC)[C@H]1CCc2nc(N)sc2C1
Tekniske data
Kemisk navn eller materiale (S)-Propyl Pramipexole
Mængde 100 mg
InChI formel InChI=1S/C13H23N3S/c1-3-7-16(8-4-2)10-5-6-11-12(9-10)17-13(14)15-11/h10H,3-9H2,1-2H3,(H2,14,15)/t10-/m0/s1
Formel vægt 253.1613