Få mere at vide
(S)-Propranolol-d7 Hydrochloride, TRC
Artikelnummer. 30421476
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30421476 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30421476 Leverandør Toronto Research Chemicals Leverandørnr. TRCP83179725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-Propranolol-d7 Hydrochloride
CAS 1346617-12-6
Molekylær formel C16 H14 D7 N O2 . HCl
Mængde 25 mg
Synonym (2S)-1-[(1-Methylethyl-d7)amino]-3-(1-naphthalenyloxy)-2-propanol Hydrochloride, S-(-)-1-(Isopropyl-d7)amino-3-(1-naphthoxy)-2-propanol Hydrochloride, (-)-Propranolol-d7 Hydrochloride, (S)-(-)-Propranolol-d7 Hydrochloride, L-(-)-Propranolol-d7 Hydrochloride, l-Propranolol-d7 Chlorhydrate, l-Propranolol-d7 Hydrochloride
InChI formel InChI=1S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18H,10-11H2,1-2H3;1H/t14-;/m0./s1/i1D3,2D3,12D;
SMIL O[C@H](COC1=CC=CC2=C1C=CC=C2)CNC(C([2H])([2H])[2H])(C([2H])([2H])[2H])[2H].Cl
IUPAC navn (S)-1-(naphthalen-1-yloxy)-3-((propan-2-yl-d7)amino)propan-2-ol hydrochloride
Molekylvægt (g/mol) 302.85
Formel vægt 302.18