Få mere at vide
(S)-Pramipexole (N-Propyl-3,3,3-d3) Dihydrochloride, TRC
Artikelnummer. 30415285
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30415285 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30415285 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7007571MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-Pramipexole (N-Propyl-3,3,3-d3) Dihydrochloride
CAS 1432230-10-8
Molekylær formel C10 D3 H14 N3 S
Mængde 1 mg
Synonym (S)-4,5,6,7-Tetrahydro-N6-(propyl-d3)-2,6-benzothiazolediamine Dihydrochloride
InChI formel InChI=1S/C10H17N3S/c1-2-5-12-7-3-4-8-9(6-7)14-10(11)13-8/h7,12H,2-6H2,1H3,(H2,11,13)/t7-/m0/s1/i1D3
SMIL [2H]C([2H])([2H])CCN[C@H]1CCc2nc(N)sc2C1
IUPAC navn (6S)-6-N-(3,3,3-trideuteriopropyl)-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine
Molekylvægt (g/mol) 214.346
Formel vægt 214.133