Få mere at vide
(S)-Pramipexole-d7 2HCl (N-propyl-d7), CDN
Artikelnummer. 30314864
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30314864 0.01 g
30313833 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314864 Leverandør CDN Leverandørnr. CDND74990.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pramipexole-D7 (N-propyl-D7) Dihydrochloride
CAS 2702798-58-9
Molekylær formel C10 D7 H10 N3 S . 2 Cl H
Mængde 0.01 g
Synonym (6 S)-N'-(1,1,2,2,3,3,3-Heptadeuteriopropyl)-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine dihydrochloride, (6 S)-4,5,6,7-Tetrahydro-N6-(propyl-d7)-2,6-benzothiazolediamine hydrochloride (1:2), Pramipexole-d7 dihydrochloride, (S)-Pramipexole (N-propyl-1,1,2,2,3,3,3-d7) dihydrochloride, Pramipexole-D7 (N-propyl-D7) dihydrochloride
InChI formel InChI=1 S/C10H17N3S0.2 ClH/c1-2-5-12-7-3-4-8-9(6-7)14-10(11)13-8;;/h7,12 H,2-6H2,1H3,(H2,11,13);2*1 H/t7-;;/m0../s1/i1D3,2D2,5D2;;
SMIL Cl.Cl.[2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N[C@H]1 CCc2nc(N)sc2C1
IUPAC navn (6 S)-6-N-(1,1,2,2,3,3,3-heptadeuteriopropyl)-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine;dihydrochloride
Molekylvægt (g/mol) 291.2921
Formel vægt 290.1116 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity