Få mere at vide
(S)-Pramipexole-d5 Dihydrochloride, TRC
Artikelnummer. 30421850
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30421850 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30421850 Leverandør Toronto Research Chemicals Leverandørnr. TRCP70075810MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-Pramipexole-(N-Propyl-2,2,3,3,3-d5) Dihydrochloride
CAS 1217601-58-5
Renhedsgrad noter HPLC
Molekylær formel C10H14D5Cl2N3S
Mængde 10 mg
Synonym (S)-4,5,6,7-Tetrahydro-N6-propyl-d5-2,6-benzothiazolediamine Dihydrochloride
InChI formel InChI=1S/C10H17N3S.2ClH/c1-2-5-12-7-3-4-8-9(6-7)14-10(11)13-8;;/h7,12H,2-6H2,1H3,(H2,11,13);2*1H/t7-;;/m0../s1/i1D3,2D2;;
SMIL NC(S1)=NC2=C1C[C@@H](NCC([2H])([2H])C([2H])([2H])[2H])CC2
Molekylvægt (g/mol) 289.28
Formel vægt 288.0991
Procent renhed >95