Få mere at vide
S-Phenylmercapturic Acid-d5, TRC
Artikelnummer. 30414383
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30414383 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30414383 Leverandør Toronto Research Chemicals Leverandørnr. TRCP3356025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale S-Phenylmercapturic Acid-d5
CAS 1331906-27-4
Renhedsgrad noter HPLC
Molekylær formel C11H8D5NO3S
Mængde 5 mg
Synonym N-Acetyl-S-phenyl-D-cysteine-d5, N-Acetyl-3-(phenylthio)alanine-d5, NSC 17197-d5, SPMA-d5
InChI formel InChI=1S/C11H13NO3S/c1-8(13)12-10(11(14)15)7-16-9-5-3-2-4-6-9/h2-6,10H,7H2,1H3,(H,12,13)(H,14,15)/t10-/m1/s1/i2D,3D,4D,5D,6D
SMIL [2H]C1=C([2H])C([2H])=C([2H])C(SC[C@@H](NC(C)=O)C(O)=O)=C1[2H]
IUPAC navn (2S)-2-acetamido-3-(2,3,4,5,6-pentadeuteriophenyl)sulfanylpropanoic acid
Molekylvægt (g/mol) 244.32
Formel vægt 244.093
Procent renhed >95