Få mere at vide
(S)-PA 824-d4 (Pretomanid-d4), TRC
Artikelnummer. 30417021
Skift visning
Mængde:
0.5 mg
2.5 mg
5 mg
Pakningsstørrelse:
0.5mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30417021 5 mg -
30414077 0.5 mg 0.5mg
30413979 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30417021 Leverandør Toronto Research Chemicals Leverandørnr. TRCP1225025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1346617-34-2
Molekylær formel C14H8D4F3N3O5
Molekylvægt (g/mol) 363.28
Synonym (6S)-6,7-Dihydro-2-nitro-6-[[4-(trifluoromethoxy)phenyl]methoxy]-5H-imidazo[2,1-b][1,3]oxazine-d4, (S)-6,7-Dihydro-2-nitro-6-[[4-(trifluoromethoxy)phenyl]methoxy]-5H-imidazo[2,1-b][1,3]oxazine-d4, PA 824-d4
SMIL FC(F)(F)OC(C=C1)=CC=C1C([2H])([2H])O[C@H](CO2)C([2H])([2H])N3C2=NC([N+]([O-])=O)=C3
Tekniske data
Kemisk navn eller materiale (S)-PA 824-d4 (Pretomanid-d4)
Renhedsgrad noter HPLC
Mængde 5 mg
InChI formel InChI=1S/C14H12F3N3O5/c15-14(16,17)25-10-3-1-9(2-4-10)7-23-11-5-19-6-12(20(21)22)18-13(19)24-8-11/h1-4,6,11H,5,7-8H2/t11-/m0/s1/i5D2,7D2
Formel vægt 363.098
Procent renhed >95%