Få mere at vide
(S)-Norfluoxetine-d5 (Phenyl-d5), TRC
Artikelnummer. 30410753
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30410753 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30410753 Leverandør Toronto Research Chemicals Leverandørnr. TRCN68109710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-Norfluoxetine-d5 (Phenyl-d5)
CAS 1217770-71-2
Molekylær formel C16H11D5F3NO
Mængde 10 mg
Synonym (γS)-γ-[4-(Trifluoromethyl)phenoxy](benzene-d5)propanamine, (S)-3-(4-Trifluoromethylphenoxy)-3-(phenyl-d5)propylamine, Seproxetine-d5
InChI formel InChI=1S/C16H16F3NO/c17-16(18,19)13-6-8-14(9-7-13)21-15(10-11-20)12-4-2-1-3-5-12/h1-9,15H,10-11,20H2/t15-/m0/s1/i1D,2D,3D,4D,5D
SMIL NCC[C@H](OC1=CC=C(C(F)(F)F)C=C1)C2=C([2H])C([2H])=C([2H])C([2H])=C2[2H]
Molekylvægt (g/mol) 300.33
Formel vægt 300.1498