Få mere at vide
S-Nitroso-N-acetyl-D,L-penicillamine, TRC
Artikelnummer. 30474871
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30474871 50 mg
30477129 25 mg
30476505 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30474871 Leverandør Toronto Research Chemicals Leverandørnr. TRCN52250050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 67776-06-1
Molekylær formel C7 H12 N2 O4 S
Molekylvægt (g/mol) 220.25
Synonym Valine, N-acetyl-3-(nitrosothio)- (9CI, ACI), DL-Valine, N-acetyl-3-(nitrosothio)- (ZCI), N-Acetyl-3-(nitrosothio)valine (ACI), 2-Acetamido-3-methyl-3-(nitrosothio)butanoic acid, N-Acetyl-S-nitroso-DL-penicillamine, N-Acetyl-S-nitrosopenicillamine, S-Nitroso-N-acetyl-DL-penicillamine, S-Nitrosoacetylpenicillamine, SNAP, SNAP (amino acid), 2-Acetamido-3-methyl-3-nitrososulfanyl-butanoic acid, N-Acetyl-3-(nitrosothio)-DL-valine
IUPAC navn 2-acetamido-3-methyl-3-nitrososulfanylbutanoic acid
SMIL CC(=O)NC(C(=O)O)C(C)(C)SN=O
Tekniske data
Kemisk navn eller materiale S-Nitroso-N-acetyl-D,L-penicillamine
Mængde 50 mg
InChI formel InChI=1S/C7H12N2O4S/c1-4(10)8-5(6(11)12)7(2,3)14-9-13/h5H,1-3H3,(H,8,10)(H,11,12)
Formel vægt 220.0518