Få mere at vide
(S)-N-Nitroso Anatabine-d4, TRC
Artikelnummer. 30465253
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30465253 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30465253 Leverandør Toronto Research Chemicals Leverandørnr. TRCN5247475MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-N-Nitrosoanatabine 2,4,5,6 D4
CAS 1426174-82-4
Renhedsgrad noter HPLC
Molekylær formel C10 2H4 H7 N3 O
Mængde 5 mg
Synonym (S)-1,2,3,6-Tetrahydro-1-nitroso-2,3'-bipyridine-2',4',5',6'-d4, (S)-N-Nitrosoanatabine D4 (pyridinyl D4), (S)-N-Nitroso Anatabine-2,4,5,6-d4, N-Nitrosoanatabine 2,4,5,6 D4
InChI formel InChI=1S/C10H11N3O/c14-12-13-7-2-1-5-10(13)9-4-3-6-11-8-9/h1-4,6,8,10H,5,7H2/t10-/m0/s1/i3D,4D,6D,8D
SMIL [2H]c1nc([2H])c([C@@H]2CC=CCN2N=O)c([2H])c1[2H]
IUPAC navn 2,3,4,6-tetradeuterio-5-[(2S)-1-nitroso-3,6-dihydro-2H-pyridin-2-yl]pyridine
Molekylvægt (g/mol) 193.24
Formel vægt 193.1153
Procent renhed >95