Få mere at vide
S,N,N’-Trimethylisothiouronium Iodide, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N,N'-S-Trimethylisothiouronium Iodide |
| CAS | 6966-83-2 |
| Molekylær formel | C4H11lN2S |
| Mængde | 5 mg |
| Synonym | Carbamimidothioic acid','N,N'-dimethyl-','methyl ester','monohydriodide (9CI), Pseudourea','1,2,3-trimethyl-2-thio-','monohydriodide (8CI), 1,2,3-Trimethyl-2-thiopseudourea hydriodide, N,N1,S-Trimethylisothiourea hydroiodide, N,N',S-Trimethylisothiouronium iodide, N,N',S-Trimethylthiopseudourea hydriodide, N,N'-Dimethyl-S-methylisothiouronium iodide, S,N,N'-Trimethylisothiouronium iodide |
| InChI formel | InChI=1S/C4H10N2S.HI/c1-5-4(6-2)7-3;/h1-3H3,(H,5,6);1H |
| SMIL | I.CN\C(=N/C)\SC |
| IUPAC navn | methyl N,N'-dimethylcarbamimidothioate;hydroiodide |
| Molekylvægt (g/mol) | 246.11 |
| Formel vægt | 245.9688 |