Få mere at vide
S,N,N'-Trimethylisothiouronium-d9 Iodide, TRC
Artikelnummer. 30449655
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30449655 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30449655 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7963472.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale S,N,N'-Trimethylisothiouronium-d9 Iodide
CAS 1331643-02-7
Molekylær formel C4H2D9IN2S
Mængde 2.5 mg
Synonym N,N,S-Trimethylthiopseudourea-d9 Hydriodide, 2,3,3-Trimethyl-2-thio-pseudourea-d9 Monohydriodide, N,N-Dimethyl-carbamimidothioic Acid-d9 Methyl Ester Monohydriodide
InChI formel InChI=1S/C4H10N2S.HI/c1-5-4(6-2)7-3;/h1-3H3,(H,5,6);1H/i1D3,2D3,3D3;
SMIL I.[2H]C([2H])([2H])N\C(=N\C([2H])([2H])[2H])\SC([2H])([2H])[2H]
IUPAC navn trideuteriomethyl N,N'-bis(trideuteriomethyl)carbamimidothioate;hydroiodide
Molekylvægt (g/mol) 255.17
Formel vægt 255.0253