Få mere at vide
(S)-N-Methyl-d5 Pregabalin, TRC
Artikelnummer. 30474155
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30474155 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30474155 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3260171MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-N-Methyl-d5 Pregabalin
CAS 1246820-67-6
Molekylær formel C9H14D5NO2
Mængde 1 mg
Synonym (3S)-3-[N-(Methyl-d5)aminomethyl)-5-methylhexanoic Acid
InChI formel InChI=1S/C9H19NO2/c1-7(2)4-8(6-10-3)5-9(11)12/h7-8,10H,4-6H2,1-3H3,(H,11,12)/t8-/m0/s1/i3D3,5D2
SMIL [2H]C([2H])([2H])NC[C@@H](CC(C)C)C([2H])([2H])C(=O)O
Molekylvægt (g/mol) 178.28