Få mere at vide
(S)-N-Desethyl Oxybutynin Hydrochloride, TRC
Artikelnummer. 30135159
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135159 5 mg
30004361 1 mg
30005375 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135159 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2894705MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 181647-14-3
Molekylær formel C20 H27 N O3 . Cl H
Molekylvægt (g/mol) 365.89
Synonym Benzeneacetic acid, α-cyclohexyl-α-hydroxy-, 4-(ethylamino)-2-butyn-1-yl ester, hydrochloride (1:1), (αS)- (ACI), Benzeneacetic acid, α-cyclohexyl-α-hydroxy-, 4-(ethylamino)-2-butynyl ester, hydrochloride, (S)- (9CI), (S)-4-(Ethylamino)but-2-yn-1-yl 2-cyclohexyl-2-hydroxy-2-phenylacetate hydrochloride, (αS)-α-Cyclohexyl-α-hydroxybenzeneacetic acid 4-(ethylamino)-2-butyn-1-yl ester hydrochloride, (S)-N-Desethyloxybutynin hydrochloride, (S)-Desethyloxybutynin hydrochloride, N-Desethyl (S)-Oxybutynin hydrochloride, (S)-Desethyl Oxybutynin hydrochloride
IUPAC navn 4-(ethylamino)but-2-ynyl (2S)-2-cyclohexyl-2-hydroxy-2-phenylacetate;hydrochloride
SMIL Cl.CCNCC#CCOC(=O)[C@](O)(C1CCCCC1)c2ccccc2
Tekniske data
Kemisk navn eller materiale (S)-N-Desethyl Oxybutynin Hydrochloride
Mængde 5 mg
InChI formel InChI=1S/C20H27NO3.ClH/c1-2-21-15-9-10-16-24-19(22)20(23,17-11-5-3-6-12-17)18-13-7-4-8-14-18;/h3,5-6,11-12,18,21,23H,2,4,7-8,13-16H2,1H3;1H/t20-;/m1./s1
Formel vægt 365.1758