Få mere at vide
S(+)-N-(2-(2-Thienyl)ethyl)-2-chlorophenyl Glycine Methyl Ester Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (S)-(+)-N-(2-(2-Thienyl)ethyl)-2-chlorophenyl Glycine methyl ester hydrochloride |
| CAS | 141109-19-5 |
| Molekylær formel | C15 H16 Cl N O2 S . Cl H |
| Mængde | 1 g |
| Synonym | Benzeneacetic acid','2-chloro-α-[[2-(2-thienyl)ethyl]amino]-','methyl ester','hydrochloride (1:1)','(αS)-, Benzeneacetic acid','2-chloro-α-[[2-(2-thienyl)ethyl]amino]-','methyl ester','hydrochloride','(S)-, Benzeneacetic acid','2-chloro-α-[[2-(2-thienyl)ethyl]amino]-','methyl ester','hydrochloride','(αS)- (9CI), (+)-(S)-Methyl α-[[2-(2-thienyl)ethyl]amino]-α-(2-chlorophenyl)acetate hydrochloride, (+)-Methyl α-(2-thienylethamino)-α-(2-chlorophenyl)acetate hydrochloride, (S)-(+)-Methyl α-[[2-(thien-2-yl)ethyl]amino]-α-(2-chlorophenyl)acetate hydrochloride |
| InChI formel | InChI=1S/C15H16ClNO2S.ClH/c1-19-15(18)14(12-6-2-3-7-13(12)16)17-9-8-11-5-4-10-20-11;/h2-7,10,14,17H,8-9H2,1H3;1H/t14-;/m0./s1 |
| SMIL | Cl.COC(=O)[C@@H](NCCc1cccs1)c2ccccc2Cl |
| IUPAC navn | methyl (2S)-2-(2-chlorophenyl)-2-(2-thiophen-2-ylethylamino)acetate;hydrochloride |
| Molekylvægt (g/mol) | 346.27 |
| Formel vægt | 345.0357 |