Få mere at vide
(S)-Metoprolol-d7, TRC
Artikelnummer. 30481306
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30481306 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30481306 Leverandør Toronto Research Chemicals Leverandørnr. TRCM33878210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-Metoprolol-d7
CAS 1292906-91-2
Molekylær formel C15H18D7NO3
Mængde 10 mg
Synonym (2S)-1-[4-(2-Methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]-2-propanol-d7
InChI formel InChI=1S/C15H25NO3/c1-12(2)16-10-14(17)11-19-15-6-4-13(5-7-15)8-9-18-3/h4-7,12,14,16-17H,8-11H2,1-3H3/t14-/m0/s1/i1D3,2D3,12D
SMIL COCCC(C=C1)=CC=C1OC[C@@H](O)CNC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn (S)-1-(4-(2-methoxyethyl)phenoxy)-3-((propan-2-yl-d7)amino)propan-2-ol
Molekylvægt (g/mol) 274.41
Formel vægt 274.23