Få mere at vide
S-Methyldihydroziprasidone, TRC
Artikelnummer. 30475250
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30475250 25 mg
30465689 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30475250 Leverandør Toronto Research Chemicals Leverandørnr. TRCM30278525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 194280-91-6
Molekylær formel C22H25ClN4OS
Molekylvægt (g/mol) 428.98
Synonym 6-Chloro-1,3-dihydro-5-[2-[4-[imino[2-(methylthio)phenyl]methyl]-1-piperazinyl]ethyl]-2H-indol-2-one, 1-[2-(6-Chloro-2,3-dihydro-2-oxo-1H-indol-5-yl)ethyl]-4-[imino[2-(methylthio)phenyl]methyl]-piperazine
IUPAC navn 6-chloro-5-[2-[4-(2-methylsulfanylbenzenecarboximidoyl)piperazin-1-yl]ethyl]-1,3-dihydroindol-2-one
SMIL O=C1NC2=CC(Cl)=C(CCN3CCN(C(C4=C(SC)C=CC=C4)=N)CC3)C=C2C1
Tekniske data
Kemisk navn eller materiale S-Methyldihydroziprasidone
Mængde 25 mg
InChI formel InChI=1S/C22H25ClN4OS/c1-29-20-5-3-2-4-17(20)22(24)27-10-8-26(9-11-27)7-6-15-12-16-13-21(28)25-19(16)14-18(15)23/h2-5,12,14,24H,6-11,13H2,1H3,(H,25,28)
Formel vægt 428.1438