Få mere at vide
S-Methyl Fenitrothion, TRC
Artikelnummer. 30135136
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135136 500 mg
30135070 250 mg
30012036 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135136 Leverandør Toronto Research Chemicals Leverandørnr. TRCM305130500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3344-14-7
Molekylær formel C9H12NO5PS
Molekylvægt (g/mol) 277.23
Synonym O,S-Dimethyl O-(3-methyl-4-nitrophenyl) Ester Phosphorothioic Acid, 8062HC, Isosumithion, Sumithion S-isomer, O,S-Dimethyl O-(3-Methyl-4-nitrophenyl) Phosphorothioate
IUPAC navn 4-[methoxy(methylsulfanyl)phosphoryl]oxy-2-methyl-1-nitrobenzene
SMIL O=P(OC)(SC)OC1=CC=C([N+]([O-])=O)C(C)=C1
Tekniske data
Kemisk navn eller materiale S-Methyl Fenitrothion
Mængde 500 mg
InChI formel InChI=1S/C9H12NO5PS/c1-7-6-8(4-5-9(7)10(11)12)15-16(13,14-2)17-3/h4-6H,1-3H3
Formel vægt 277.0174