Få mere at vide
S-Methyl Ergothioneine, TRC
Artikelnummer. 30481330
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30481330 10 mg
30480260 2.5 mg
30471148 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30481330 Leverandør Toronto Research Chemicals Leverandørnr. TRCM30461010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 176982-20-0
Molekylær formel C10H17N3O2S
Molekylvægt (g/mol) 243.33
Synonym (alphaS)-alpha-Carboxy-N,N,N-trimethyl-2-(methylthio)-1H-imidazole-5-ethanaminium Inner Salt, (alphaS)-alpha-Carboxy-N,N,N-trimethyl-2-(methylthio)-1H-imidazole-4-ethanaminium Inner Salt
IUPAC navn 3-(2-methylsulfanyl-1H-imidazol-5-yl)-2-(trimethylazaniumyl)propanoate
SMIL CSc1ncc(CC(C(=O)[O-])[N+](C)(C)C)[nH]1
Tekniske data
Kemisk navn eller materiale S-Methyl Ergothioneine
Mængde 10 mg
InChI formel InChI=1S/C10H17N3O2S/c1-13(2,3)8(9(14)15)5-7-6-11-10(12-7)16-4/h6,8H,5H2,1-4H3,(H-,11,12,14,15)
Formel vægt 243.1041