Få mere at vide
(S)-(-)-Malic-2,3,3-d3 Acid, CDN
Artikelnummer. 30321492
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321492 0.005 g
30309382 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321492 Leverandør CDN Leverandørnr. CDND66260.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Malic Acid-2,3,3-D3
CAS 59652-74-3
Molekylær formel C4 D3 H3 O5
Mængde 0.005 g
Synonym Butanedioic-d3 acid, hydroxy-, - (9 CI), (2 S)-2-Hydroxybutanedioic-2,3,3-d2 acid, L-Malic acid-2,3,3-d3, (S)-(-)-Malic-2,3,3-d3 Acid
InChI formel InChI=1 S/C4H6O5/c5-2(4(8)9)1-3(6)7/h2,5 H,1H2,(H,6,7)(H,8,9)/t2-/m0/s1/i1D2,2 D
SMIL [2 H]C([2 H])(C(=O)O)[C@]([2 H])(O)C(=O)O
IUPAC navn (3 S)-2,2,3-trideuterio-3-hydroxybutanedioic acid
Molekylvægt (g/mol) 137.1059
Formel vægt 137.0404 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity