missing translation for 'onlineSavingsMsg'
Få mere at vide

(S)-Lisinopril Dimer, TRC

Artikelnummer. 30479361
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479361 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30479361 Leverandør Toronto Research Chemicals Leverandørnr. TRCL46901010MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Lisinopril Dimer
CAS 1356839-89-8
Molekylær formel C42 H60 N6 O9
Mængde 10 mg
Synonym Lisinopril Dimer, (S)-Lisinopril Dimer, (2S)-1-[(2S)-6-Amino-2-[[(2S)-1-[[[(5S)-5-[[(1S)-1-carboxy-3-phenylpropyl]amino]-6-[(2S)-2-carboxypyrrolidin-1-yl]-6-oxohexyl]amino]carbonyl]-3-phenylpropyl]amino]hexanoyl]pyrrolidine-2-carboxylic Acid, Lisinopril Dihydrate Imp. G (EP)
InChI formel InChI=1S/C42H60N6O9/c43-25-9-7-17-32(38(50)47-27-11-19-35(47)41(54)55)45-31(23-21-29-13-3-1-4-14-29)37(49)44-26-10-8-18-33(39(51)48-28-12-20-36(48)42(56)57)46-34(40(52)53)24-22-30-15-5-2-6-16-30/h1-6,13-16,31-36,45-46H,7-12,17-28,43H2,(H,44,49)(H,52,53)(H,54,55)(H,56,57)/t31-,32-,33-,34-,35-,36-/m0/s1
SMIL NCCCC[C@H](N[C@@H](CCc1ccccc1)C(=O)NCCCC[C@H](N[C@@H](CCc2ccccc2)C(=O)O)C(=O)N3CCC[C@H]3C(=O)O)C(=O)N4CCC[C@H]4C(=O)O
IUPAC navn (2S)-1-[(2S)-6-amino-2-[[(2S)-1-[[(5S)-5-[[(1S)-1-carboxy-3-phenylpropyl]amino]-6-[(2S)-2-carboxypyrrolidin-1-yl]-6-oxohexyl]amino]-1-oxo-4-phenylbutan-2-yl]amino]hexanoyl]pyrrolidine-2-carboxylic acid
Molekylvægt (g/mol) 792.96
Formel vægt 792.4422
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.