Få mere at vide
(S)-Lisinopril-d5 Sodium, TRC
Artikelnummer. 30478839
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30478839 10 mg
30479360 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30478839 Leverandør Toronto Research Chemicals Leverandørnr. TRCL46898710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1356905-39-9
Molekylær formel C21H26D5N3NaO5
Molekylvægt (g/mol) 433.51
Synonym N2-[(1S)-1-Carboxy-3-(phenyl-d5)propyl]-L-lysyl-L-proline, Lisinopril-d5, (2S)-1-[(2S)-6-Amino-2-[[(1S)-1-carboxy-3-(phenyl-d5)propyl]amino]hexanoyl]pyrrolidine-2-carboxylic acid, Lisinopril-D5 (phenyl-D5)
SMIL [Na+].[2H]c1c([2H])c([2H])c(CC[C@H](N[C@@H](CCCCN)C(=O)N2CCC[C@H]2C(=O)[O-])C(=O)O)c([2H])c1[2H]
Tekniske data
Kemisk navn eller materiale (S) Lisinopril-d5
Mængde 10 mg
InChI formel InChI=1S/C21H31N3O5/c22-13-5-4-9-16(19(25)24-14-6-10-18(24)21(28)29)23-17(20(26)27)12-11-15-7-2-1-3-8-15/h1-3,7-8,16-18,23H,4-6,9-14,22H2,(H,26,27)(H,28,29)/t16-,17-,18-/m0/s1/i1D,2D,3D,7D,8D