Få mere at vide
(S)-Linezolid-d3 (acetamide-d3), CDN
Artikelnummer. 30309476
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30309476 0.005 g
30314579 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309476 Leverandør CDN Leverandørnr. CDND76630.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Linezolid-d3
CAS 1127120-38-0
Molekylær formel C16 D3 H17 F N3 O4
Mængde 0.005 g
Synonym 2,2,2-Trideuterio-N-[[(5 S)-3-(3-fluoro-4-morpholin-4-ylphenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl]acetamide, N-[[(5 S)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]acetamide-2,2,2-d3, Linezolid-D3 (acetyl-D3), (S)-Linezolid-d3, (S)-Linezolid-d3 (acetamide-d3)
InChI formel InChI=1 S/C16H20FN3O4/c1-11(21)18-9-13-10-20(16(22)24-13)12-2-3-15(14(17)8-12)19-4-6-23-7-5-19/h2-3,8,13 H,4-7,9-10H2,1H3,(H,18,21)/t13-/m0/s1/i1D3
SMIL [2 H]C([2 H])([2 H])C(=O)NC[C@H]1 CN(C(=O)O1)c2ccc(N3CCOCC3)c(F)c2
IUPAC navn 2,2,2-trideuterio-N-[[(5 S)-3-(3-fluoro-4-morpholin-4-ylphenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl]acetamide
Molekylvægt (g/mol) 340.365
Formel vægt 340.163 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity