Få mere at vide
(S)-Hydroxychloroquine(>90%), TRC
Artikelnummer. 30367115
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367115 25 mg
30361283 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30367115 Leverandør Toronto Research Chemicals Leverandørnr. TRCC37371525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 137433-24-0
Molekylær formel C18 H26 Cl N3 O
Molekylvægt (g/mol) 335.87
Synonym Ethanol, 2-[[4-[(7-chloro-4-quinolinyl)amino]pentyl]ethylamino]-, (S)-, 2-[[(4S)-4-[(7-Chloro-4-quinolinyl)amino]pentyl]ethylamino]ethanol, 2-[[(4S)-4-[(7-chloro-4-quinolinyl)amino]pentyl]ethylamino]-ethanol
IUPAC navn 2-[[(4S)-4-[(7-chloroquinolin-4-yl)amino]pentyl]-ethylamino]ethanol
SMIL CCN(CCO)CCC[C@H](C)Nc1ccnc2cc(Cl)ccc12
Tekniske data
Kemisk navn eller materiale (S)-Hydroxychloroquine
Mængde 25 mg
InChI formel InChI=1S/C18H26ClN3O/c1-3-22(11-12-23)10-4-5-14(2)21-17-8-9-20-18-13-15(19)6-7-16(17)18/h6-9,13-14,23H,3-5,10-12H2,1-2H3,(H,20,21)/t14-/m0/s1
Formel vægt 335.1764