Få mere at vide
(S)-Hydroperoxy Des(boric Acid) Bortezomib, TRC
Artikelnummer. 30136729
Skift visning
Mængde:
1 mg
2.5 mg
10 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30136729 10 mg -
30135601 2.5 mg 2.5mg
30005084 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136729 Leverandør Toronto Research Chemicals Leverandørnr. TRCH71540510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 886979-81-3
Molekylær formel C19 H24 N4 O4
Molekylvægt (g/mol) 372.42
Synonym 2-Pyrazinecarboxamide, N-[(1S)-2-[[(1S)-1-hydroperoxy-3-methylbutyl]amino]-2-oxo-1-(phenylmethyl)ethyl]-, Pyrazinecarboxamide, N-[(1S)-2-[[(1S)-1-hydroperoxy-3-methylbutyl]amino]-2-oxo-1-(phenylmethyl)ethyl]- (9CI), Bortezomib S-hydroperoxide (USP)
IUPAC navn N-[(2S)-1-[[(1S)-1-hydroperoxy-3-methylbutyl]amino]-1-oxo-3-phenylpropan-2-yl]pyrazine-2-carboxamide
SMIL CC(C)C[C@@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)c2cnccn2)OO
Tekniske data
Kemisk navn eller materiale (S)-Hydroperoxy Des(boric Acid) Bortezomib
InChI formel InChI=1S/C19H24N4O4/c1-13(2)10-17(27-26)23-18(24)15(11-14-6-4-3-5-7-14)22-19(25)16-12-20-8-9-21-16/h3-9,12-13,15,17,26H,10-11H2,1-2H3,(H,22,25)(H,23,24)/t15-,17-/m0/s1
Mængde 10 mg
Formel vægt 372.1798