Få mere at vide
S-Desethylpiperidino S-(Glycin-N-yl) Vardenafil, TRC
Artikelnummer. 30138714
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138714 10 mg
30137018 25 mg
30006731 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138714 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29097510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 448184-54-1
Molekylær formel C19H23N5O6S
Molekylvægt (g/mol) 449.48
Synonym N-[[3-(1,4-Dihydro-5-methyl-4-oxo-7-propylimidazo[5,1-f][1,2,4]triazin-2-yl)-4-ethoxyphenyl]sulfonyl]glycine, Vardenafil Glycene Impurity
SMIL O=C(O)CNS(=O)(C1=CC=C(OCC)C(C(NN2C3=C(C)N=C2CCC)=NC3=O)=C1)=O
Tekniske data
Kemisk navn eller materiale S-Desethylpiperidino S-(Glycin-N-yl) Vardenafil
Analyt- eller komponentnavne S-Desethylpiperidino S-(Glycin-N-yl) Vardenafil
Forsendelsestilstand Room Temperature
Løsningstype Neat
Industry Type Pharmaceutical