Få mere at vide
(S)-Citalopram-d6 Oxalate, TRC
Artikelnummer. 30354350
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30354350 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30354350 Leverandør Toronto Research Chemicals Leverandørnr. TRCC50501210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-Citalopram N,N-dimethyl-d6 Oxalate
CAS 1217733-09-9
Renhedsgrad noter HPLC
Molekylær formel C20 2H6 H15 F N2 O . C2 H2 O4
Mængde 10 mg
Synonym (1S)-1-[3-(Bis(trideuteriomethyl)amino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile oxalic acid, (1S)-1-[3-[Di(methyl-d3)amino]propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile 1,2-ethanedioate, (1S)-1-[3-(Bis(trideuteriomethyl)amino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile oxalate, (1S)-1-(3-[Di(methyl-d3)amino]propyl)-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile oxalate, Escitalopram-d6 oxalate, (S)-Citalopram-d6 oxalate, (S)-Citalopram-D6 oxalate (N,N-dimethyl-D6)
InChI formel InChI=1S/C20H21FN2O.C2H2O4/c1-23(2)11-3-10-20(17-5-7-18(21)8-6-17)19-9-4-15(13-22)12-16(19)14-24-20;3-1(4)2(5)6/h4-9,12H,3,10-11,14H2,1-2H3;(H,3,4)(H,5,6)/t20-;/m0./s1/i1D3,2D3;
SMIL [2H]C([2H])([2H])N(CCC[C@]1(OCc2cc(ccc12)C#N)c3ccc(F)cc3)C([2H])([2H])[2H].OC(=O)C(=O)O
IUPAC navn (1S)-1-[3-[bis(trideuteriomethyl)amino]propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;oxalic acid
Molekylvægt (g/mol) 420.46
Formel vægt 420.1968
Procent renhed >95