Få mere at vide
(S)-Chlorpheniramine Maleate Salt, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Dexchlorpheniramine Maleate |
| CAS | 2438-32-6 |
| Molekylær formel | C16 H19 Cl N2 . C4 H4 O4 |
| Mængde | 1 g |
| Synonym | Dexchlorpheniramine Maleate, Brompheniramine Maleate Imp. B (EP) as Maleate, 2-Pyridinepropanamine, γ-(4-chlorophenyl)-N,N-dimethyl-, (γS)-, (2Z)-2-butenedioate (1:1) (9CI, ACI), 2-Pyridinepropanamine, γ-(4-chlorophenyl)-N,N-dimethyl-, (S)-, (Z)-2-butenedioate (1:1) (ZCI), Pyridine, 2-[p-chloro-α-[2-(dimethylamino)ethyl]benzyl]-, maleate (1:1), (S)- (8CI), (+)-Chlorpheniramine maleate, (S)-3-(4-Chlorophenyl)-N,N-dimethyl-3-(pyridin-2-yl)propan-1-amine maleate, d-2-[p-Chloro-α-(2-dimethylaminoethyl)benzyl]pyridine bimaleate, d-Chlorpheniramine maleate, d-Chlorpheniramine maleate, Dexchloropheniramine maleate, Dexchlorpheniramine hydrogen maleate, Dexchlorpheniramine maleate, Dextrochlorpheniramine maleate, Fortamine, Isomerine, Phenamin, Phendextro, Polamin, Polaramin, Polaramine, Polaronil, S-(+)-Chlorpheniramine maleate, S-Chlorpheniramine maleate, Sensidyn |
| InChI formel | InChI=1S/C16H19ClN2.C4H4O4/c1-19(2)12-10-15(16-5-3-4-11-18-16)13-6-8-14(17)9-7-13;5-3(6)1-2-4(7)8/h3-9,11,15H,10,12H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t15-;/m0./s1 |
| SMIL | CN(C)CC[C@@H](c1ccc(Cl)cc1)c2ccccn2.OC(=O)\C=C/C(=O)O |
| IUPAC navn | (Z)-but-2-enedioic acid;(3S)-3-(4-chlorophenyl)-N,N-dimethyl-3-pyridin-2-ylpropan-1-amine |
| Molekylvægt (g/mol) | 390.86 |
| Formel vægt | 390.1346 |