Få mere at vide
(S)-(-)-Celiprolol-d9 Hydrochloride, TRC
Artikelnummer. 30366067
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366067 1 mg
30352015 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30366067 Leverandør Toronto Research Chemicals Leverandørnr. TRCC2544971MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1330266-42-6
Molekylær formel C20 D9 H24 N3 O4 . H Cl
Molekylvægt (g/mol) 425.01
Synonym N'-[3-Acetyl-4-[(2S)-3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]phenyl]-N,N-diethylurea-d9 Hydrochloride, (-)-(S)-Celiprolol-d9 Hydrochloride, (-)-Celiprolol-d9 Hydrochloride, (S)-Celiprolol-d9 Hydrochloride, l-Celiprolol-d9 Hydrochloride
IUPAC navn 3-[3-acetyl-4-[(2S)-3-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]amino]-2-hydroxypropoxy]phenyl]-1,1-diethylurea;hydrochloride
SMIL Cl.[2H]C([2H])([2H])C(NC[C@H](O)COc1ccc(NC(=O)N(CC)CC)cc1C(=O)C)(C([2H])([2H])[2H])C([2H])([2H])[2H]
Tekniske data
Kemisk navn eller materiale (S)-(-)-Celiprolol-d9 Hydrochloride
Mængde 1 mg
InChI formel InChI=1S/C20H33N3O4.ClH/c1-7-23(8-2)19(26)22-15-9-10-18(17(11-15)14(3)24)27-13-16(25)12-21-20(4,5)6;/h9-11,16,21,25H,7-8,12-13H2,1-6H3,(H,22,26);1H/t16-;/m0./s1/i4D3,5D3,6D3;
Formel vægt 424.28