Få mere at vide
(S)-Beta-Phenylalanine, TRC
Artikelnummer. 30418371
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418371 10 g
30412544 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30418371 Leverandør Toronto Research Chemicals Leverandørnr. TRCP31943010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-beta-Phenylalanine
CAS 40856-44-8
Molekylær formel C9H11NO2
Mængde 10 g
Synonym (betaS)-beta-Aminobenzenepropanoic Acid, (-)-3-Amino-3-phenylpropanoic Acid, (-)-beta-Aminobenzenepropanoic Acid, (S)-3-Amino-3-phenylpropanoic Acid, (S)-3-Amino-3-phenylpropionic Acid, (S)-beta-Phenylalanine, D-3-Amino-3-phenylpropionic Acid, (S)-beta-Aminobenzenepropanoic Acid
InChI formel InChI=1S/C9H11NO2/c10-8(6-9(11)12)7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/t8-/m0/s1
SMIL N[C@@H](CC(=O)O)c1ccccc1
IUPAC navn (3S)-3-amino-3-phenylpropanoic acid
Molekylvægt (g/mol) 165.19
Formel vægt 165.079