Få mere at vide
(S)-Baclofen, TRC
Artikelnummer. 30136360
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136360 25 mg
30134011 10 mg
30004939 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136360 Leverandør Toronto Research Chemicals Leverandørnr. TRCB10799525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 66514-99-6
Molekylær formel C10 H12 Cl N O2
Molekylvægt (g/mol) 213.66
Synonym (βS)-β-(Aminomethyl)-4-chlorobenzenepropanoic Acid, (+)-Baclofen, (S)-4-Amino-3-(4-chlorophenyl)butanoic Acid, (S)-4-Amino-3-(4-chlorophenyl)butyric Acid, (S)-Baclofen, L-(+)-Baclofen, L-Baclofen, S(+)-Baclofen, d-Baclofen;
IUPAC navn (3S)-4-amino-3-(4-chlorophenyl)butanoic acid
SMIL NC[C@@H](CC(=O)O)c1ccc(Cl)cc1
Tekniske data
Kemisk navn eller materiale (S)-Baclofen
InChI formel InChI=1S/C10H12ClNO2/c11-9-3-1-7(2-4-9)8(6-12)5-10(13)14/h1-4,8H,5-6,12H2,(H,13,14)/t8-/m1/s1
Mængde 25 mg
Formel vægt 213.0557