missing translation for 'onlineSavingsMsg'
Få mere at vide

(S)-Alpha-Terpineol, TRC

Artikelnummer. 17973735
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17973735 10 g
17902125 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Product Code. 17973735 Supplier Toronto Research Chemicals Supplier No. TRCT11749510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10482-56-1
Molekylær formel C10 H18 O
Molekylvægt (g/mol) 154.25
Synonym 3-Cyclohexene-1-methanol, α,α,4-trimethyl-, (1S)-, 3-Cyclohexene-1-methanol, α,α,4-trimethyl-, (S)-, p-Menth-1-en-8-ol, (S)-(-)- (8CI), (1S)-α,α,4-Trimethyl-3-cyclohexene-1-methanol, (-)-α-Terpineol, (S)-(-)-Terpineol, (S)-(-)-α-Terpineol, (S)-α,α,4-Trimethyl-3-cyclohexene-1-methanol, (S)-α-Terpineol, l-α-Terpineol, α-Terpineol, (-)-, (S)-alpha-Terpineol
IUPAC navn 2-[(1S)-4-methylcyclohex-3-en-1-yl]propan-2-ol
SMIL CC1=CC[C@H](CC1)C(C)(C)O

Specifications

Kemisk navn eller materiale (S)-alpha-Terpineol
Mængde 10 g
InChI formel InChI=1S/C10H18O/c1-8-4-6-9(7-5-8)10(2,3)11/h4,9,11H,5-7H2,1-3H3/t9-/m1/s1
Formel vægt 154.1358
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.