missing translation for 'onlineSavingsMsg'
Få mere at vide

(S)-7-Hydroxy Warfarin, TRC

Artikelnummer. 30406268
Change view
Click to view available options
Mængde:
1 mg
2.5 mg
25 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
30406268 25 mg -
30391473 1 mg -
30396783 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406268 Supplier Toronto Research Chemicals Supplier No. TRCH99612625MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63740-81-8
Molekylær formel C19 H16 O5
Molekylvægt (g/mol) 324.33
Synonym 2H-1-Benzopyran-2-one, 4,7-dihydroxy-3-[(1S)-3-oxo-1-phenylbutyl]-, 2H-1-Benzopyran-2-one, 4,7-dihydroxy-3-(3-oxo-1-phenylbutyl)-, (S)-, 4,7-Dihydroxy-3-[(1S)-3-oxo-1-phenylbutyl]-2H-1-benzopyran-2-one, 7-Hydroxy-S-warfarin, S-7-Hydroxywarfarin, (S)-7-Hydroxywarfarin
IUPAC navn 4,7-dihydroxy-3-[(1S)-3-oxo-1-phenylbutyl]chromen-2-one
SMIL CC(=O)C[C@H](C1=C(O)c2ccc(O)cc2OC1=O)c3ccccc3

Specifications

Kemisk navn eller materiale (S)-7-Hydroxywarfarin
Mængde 25 mg
InChI formel InChI=1S/C19H16O5/c1-11(20)9-15(12-5-3-2-4-6-12)17-18(22)14-8-7-13(21)10-16(14)24-19(17)23/h2-8,10,15,21-22H,9H2,1H3/t15-/m0/s1
Formel vægt 324.0998
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.