missing translation for 'onlineSavingsMsg'
Learn More

(S)-5-Chloro Naproxen, TRC

Product Code. 30349523
Change view
Click to view available options
Mængde:
250 mg
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
30349523 250 mg -
30358996 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30349523 Supplier Toronto Research Chemicals Supplier No. TRCC369715250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89617-86-7
Molekylær formel C14 H13 Cl O3
Molekylvægt (g/mol) 264.7
Synonym (2S)-2-(5-Chloro-6-methoxynaphthalen-2-yl)propanoic Acid, Naproxen Imp. B (EP), 5-Chloronaproxen
IUPAC navn (2S)-2-(5-chloro-6-methoxynaphthalen-2-yl)propanoic acid
SMIL COc1ccc2cc(ccc2c1Cl)[C@H](C)C(=O)O

Specifications

Kemisk navn eller materiale (2S)-2-(5-Chloro-6-methoxynaphthalen-2-yl)propanoic Acid (5-Chloronaproxen)
Mængde 250 mg
InChI formel InChI=1S/C14H13ClO3/c1-8(14(16)17)9-3-5-11-10(7-9)4-6-12(18-2)13(11)15/h3-8H,1-2H3,(H,16,17)/t8-/m0/s1
Formel vægt 264.0553
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.