Få mere at vide
S-(4-Nitrophenyl)mercapturic Acid, TRC
Artikelnummer. 30484288
Skift visning
Mængde:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30484288 50 mg
30464675 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30484288 Leverandør Toronto Research Chemicals Leverandørnr. TRCN50441050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 91088-55-0
Molekylær formel C11 H12 N2 O5 S
Molekylvægt (g/mol) 284.29
IUPAC navn (2R)-2-acetamido-3-(4-nitrophenyl)sulfanylpropanoic acid
SMIL CC(=O)N[C@@H](CSc1ccc(cc1)[N+](=O)[O-])C(=O)O
Tekniske data
Kemisk navn eller materiale S-(4-Nitrophenyl)mercapturic Acid
Mængde 50 mg
InChI formel InChI=1S/C11H12N2O5S/c1-7(14)12-10(11(15)16)6-19-9-4-2-8(3-5-9)13(17)18/h2-5,10H,6H2,1H3,(H,12,14)(H,15,16)/t10-/m0/s1
Formel vægt 284.0467